β-Alanine 98%

Supplier: Thermo Scientific Chemicals

A16665-0B A16665-0I A16665-30
CAAAA16665-0BEA 132.25 CAD
CAAAA16665-0B CAAAA16665-0I CAAAA16665-30
β-Alanine 98%
β-Alanine

Purity: 98% CAS No.: 107-95-9 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Physical Form: Cryst powder Density: 1.437 Appearance: White Odor: Odorless Melting Point: ca 205° dec. Beilstein Reference: 906793 Merck Reference: 14,205 MDL No.: MFCD00008200

Formula: C₃H₇NO₂
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: ∼ 205 °C (dec.)
Density: 1.437
Storage Temperature: Ambient
MDL Number: MFCD00008200
CAS Number: 107-95-9
EINECS: 203-536-5
Merck Index: 14,205

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR