Diphenylboric acid-2-aminoethyl ester (2-APB) 98%

Supplier: Thermo Scientific Chemicals
Warning

A16606-06 A16606-14
CAAAA16606-06EA 123.24 CAD
CAAAA16606-06 CAAAA16606-14
Diphenylboric acid-2-aminoethyl ester (2-APB) 98%
Diphenylboric acid 2-aminoethyl ester (2-APB)

Purity: 98% CAS No.: 524-95-8 Molecular Formula: C14H16BNO Formula Weight: 225.10 Melting Point: 187-190° Beilstein Reference: 2942273

Formula: C₁₄H₁₆BNO
Melting Pt: 187…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00014823
CAS Number: 524-95-8
EINECS: 208-366-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR