(3,4-Difluorophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Difluorophenylacetic acid

A16584-06
CAAAAA16584-14EA 853.53 CAD
CAAAAA16584-14 CAAAA16584-03 CAAAA16584-06
(3,4-Difluorophenyl)acetic acid 98%
(3,4-Difluorophenyl)acetic acid

Purity: 98% CAS No.: 658-93-5 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Flash Point: 110°(230°F) Melting Point: 48-50° Beilstein Reference: 2577768 MDL No.: MFCD00010002

Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259
Melting Pt: 48…50 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010002
CAS Number: 658-93-5
EINECS: 211-527-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR