Brucine dihydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A16251-09 A16251-18
CAAAA16251-09EA 69.52 CAD
CAAAA16251-09 CAAAA16251-18
Brucine dihydrate 98%
Brucine dihydrate

Brucine dihydrate is used as a resolving agent for chiral acids.


Purity: 98% CAS No.: 5892-11-5 Molecular Formula: C23H26N2O4.2H2O Formula Weight: 430.50 (394.47anhy) Melting Point: 176-178° UN No.: 1570 MDL No.: MFCD00149384

Formula: C₂₃H₂₆N₂O₄·2H₂O
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00149384
CAS Number: 5892-11-5
UN: 1570
ADR: 6.1,I

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR