L(+)-Lysine dihydrochloride 99%

Supplier: Thermo Scientific Chemicals

A16127-14 A16127-22
CAAAA16127-14EA 49.3 CAD
CAAAA16127-14 CAAAA16127-22
L(+)-Lysine dihydrochloride 99%
L(+)-Lysine dihydrochloride

Purity: 99% CAS No.: 657-26-1 Molecular Formula: C6H14N2O2.2HCl Formula Weight: 219.11 Melting Point: 202-206° Beilstein Reference: 4539715 Merck Reference: 14,5636 MDL No.: MFCD00066389

Formula: C₆H₁₆Cl₂N₂O₂
Melting Pt: 202…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00066389
CAS Number: 657-26-1
EINECS: 211-518-3
Merck Index: 14,5636

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR