3-Methyl-2-buten-1-ol 98+%

Supplier: Thermo Scientific Chemicals
Danger

A16089-14 A16089-22 A16089-36
CAAAA16089-14EA 48.82 CAD
CAAAA16089-14 CAAAA16089-22 CAAAA16089-36
3-Methyl-2-buten-1-ol 98+%
3-Methyl-2-buten-1-ol

Purity: 98+% CAS No.: 556-82-1 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 43°(109°F) Refractive Index: 1.4440 Boiling Point: 139-140° Beilstein Reference: 1633479 UN No.: 1987 MDL No.: MFCD00002916

Formula: C₅H₁₀O
Boiling Pt: 139…140 °C
Melting Pt: –86 °C
Density: 0.860
Flash Pt: 43 °C (109 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002916
CAS Number: 556-82-1
EINECS: 209-141-4
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR