4-tert-Butyl-1-bromobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

A15924-36 A15924-14 A15924-22
CAAAAA15924-36EA 1000.14 CAD
CAAAAA15924-36 CAAAA15924-14 CAAAA15924-22
4-tert-Butyl-1-bromobenzene 97%
4-tert-Butyl-1-bromobenzene

Purity: 97% CAS No.: 3972-65-4 Molecular Formula: C10H13Br Formula Weight: 213.12 Flash Point: 97°(206°F) Refractive Index: 1.5330 Melting Point: 15-16° Boiling Point: 231-232° Beilstein Reference: 1859117 MDL No.: MFCD00000108

Formula: C₁₀H₁₃Br
Boiling Pt: 231…232 °C
Melting Pt: 15…16 °C
Density: 1,229
Flash Pt: 97 °C(206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000108
CAS Number: 3972-65-4
EINECS: 223-599-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR