2-Ethyl-4-methylimidazole 96%

Supplier: Thermo Scientific Chemicals
Danger

A15798-14 A15798-22
CAAAA15798-14EA 63.2 CAD
CAAAA15798-14 CAAAA15798-22
2-Ethyl-4-methylimidazole 96%
2-Ethyl-4-methylimidazole

Purity: 96% CAS No.: 931-36-2 Molecular Formula: C6H10N2 Formula Weight: 110.16 Flash Point: 137°(278°F) Refractive Index: 1.5 00 Melting Point: 45-49° Boiling Point: 292-295° Beilstein Reference: 1711 MDL No.: MFCD00005193

Formula: C₆H₁₀N₂
Boiling Pt: 292…295 °C
Melting Pt: 45…49 °C
Density: 0.973
Flash Pt: 137 °C(278 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005193
CAS Number: 931-36-2
EINECS: 213-234-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR