4-Nitrobenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning

A15742-36 A15742-14 A15742-22
CAAAAA15742-36EA 676.24 CAD
CAAAAA15742-36 CAAAA15742-14 CAAAA15742-22
4-Nitrobenzyl alcohol 99%
4-Nitrobenzyl alcohol

Purity: 99% CAS No.: 619-73-8 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 180°(356°F) Melting Point: 92-95° Boiling Point: 185°/12mm Beilstein Reference: 1424026 MDL No.: MFCD00007376

Formula: C₇H₇NO₃
Boiling Pt: 185 °C/12 mm
Melting Pt: 92…95 °C
Flash Pt: 180 °C(356 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007376
CAS Number: 619-73-8
EINECS: 210-611-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR