2-Amino-5-chlorobenzophenone 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15720-0E A15720-22 A15720-36
CAAAA15720-0EEA 707.84 CAD
CAAAA15720-0E CAAAA15720-22 CAAAA15720-36
2-Amino-5-chlorobenzophenone 98+%
2-Amino-5-chlorobenzophenone

Purity: 98+% CAS No.: 719-59-5 Molecular Formula: C13H10ClNO Formula Weight: 231.68 Flash Point: 211°(412°F) Melting Point: 96-99° Beilstein Reference: 475640 MDL No.: MFCD00007839

Formula: C₁₃H₁₀ClNO
Boiling Pt: 424 °C (1013 hPa)
Melting Pt: 96…99 °C
Density: 1,33
Flash Pt: 211 °C(412 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007839
CAS Number: 719-59-5
EINECS: 211-949-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR