Ethyl malonyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

A15616-06 A15616-14 A15616-22
CAAAA15616-06EA 109.81 CAD
CAAAA15616-06 CAAAA15616-14 CAAAA15616-22
Ethyl malonyl chloride 95%
Ethyl malonyl chloride

Purity: 95% CAS No.: 36239-09-5 Molecular Formula: C5H7ClO3 Formula Weight: 150.56 Storage Temperature: 2°C to 8°C Flash Point: 67°(152°F) Refractive Index: 1.4290 Boiling Point: 83°/22mm Beilstein Reference: 636215 UN No.: 3265 MDL No.: MFCD00000736

Formula: C₅H₇ClO₃
Boiling Pt: 83 °C/22 mm
Density: 1.189
Flash Pt: 67 °C (152 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000736
CAS Number: 36239-09-5
EINECS: 252-934-5
UN: 3265
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR