Ethyl oleate 70%, tech.

Supplier: Thermo Scientific Chemicals

Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester

A15601-0B A15601-0I A15601-30
CAAAA15601-0BEA 162.74 CAD
CAAAA15601-0B CAAAA15601-0I CAAAA15601-30
Ethyl oleate 70%, tech.
Ethyl oleate

Purity: 70% CAS No.: 111-62-6 Molecular Formula: C20H38O2 Formula Weight: 310.53 Flash Point: 110°(230°F) Refractive Index: 1.451 Melting Point: -32° Boiling Point: 205-208° Beilstein Reference: 1727318 MDL No.: MFCD00009579

Formula: C₂₀H₃₈O₂
MW: 310.52 g/mol
Boiling Pt: 205…208 °C
Melting Pt: –32 °C
Density: 0.87
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009579
CAS Number: 111-62-6
EINECS: 203-889-5
Merck Index: 12,06965

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR