2-Nitro-4,5-xylidine 97%

Supplier: Thermo Scientific Chemicals
Warning

A15556-06 A15556-14 A15556-22
CAAAA15556-06EA 65.1 CAD
CAAAA15556-06 CAAAA15556-14 CAAAA15556-22
2-Nitro-4,5-xylidine 97%
2-Nitro-4,5-xylidine

Purity: 97% CAS No.: 6972-71-0 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Melting Point: 138-143° Beilstein Reference: 2209637 UN No.: 2811 MDL No.: MFCD00007811

Formula: C₈H₁₀N₂O₂
MW: 166.18
Melting Pt: 143
Density: 1.398
Storage Temperature: Ambient
MDL Number: MFCD00007811
CAS Number: 6972-71-0
EINECS: 230-211-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR