4'-(Trifluoromethyl)acetophenone 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-[4-(Trifluoromethyl)phenyl]ethan-1-one, 4-(Trifluoromethyl)acetophenone, p-(Trifluoromethyl)acetophenone

A15529-06 A15529-14
CAAAA15529-06EA 92.27 CAD
CAAAA15529-06 CAAAA15529-14 CAAAA15529-22
4'-(Trifluoromethyl)acetophenone 98+%
4'-(Trifluoromethyl)acetophenone

Purity: 98+% CAS No.: 709-63-7 Molecular Formula: C9H7F3O Formula Weight: 188.15 Flash Point: 84°(183°F) Refractive Index: 1.4620 Odor: Characteristic Melting Point: 29-35° Boiling Point: 83-85°/9mm Beilstein Reference: 1870425 MDL No.: MFCD00000401

Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 83…85 °C (9 mmHg)
Melting Pt: 29…35 °C
Density: 0.924
Flash Pt: 84 °C (183 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000401
CAS Number: 709-63-7
EINECS: 211-913-0
UN: 1325
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR