Phenetole 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15501-0F A15501-AE A15501-AP
CAAAA15501-0FEA 420.28 CAD
CAAAA15501-0F CAAAA15501-AE CAAAA15501-AP
Phenetole 98+%
Phenetole

Purity: 98+% CAS No.: 103-73-1 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 57°(134°F) Refractive Index: 1.5070 Odor: Aromatic Melting Point: -30° Boiling Point: 169-170° Beilstein Reference: 636270 Merck Reference: 14,7229 UN No.: 3271 MDL No.: MFCD00009090

Formula: C₆H₅OC₂H₅
Boiling Pt: 169…170 °C
Melting Pt: –30 °C
Density: 0.967
Flash Pt: 57 °C (134 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009090
CAS Number: 103-73-1
EINECS: 203-139-7
UN: 3271
ADR: 3,III
Merck Index: 14,7229

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR