Diethyl malonate 99%

Supplier: Thermo Scientific Chemicals
Warning

A15468-0C A15468-0E A15468-36
CAAAA15468-0CEA 597.24 CAD
CAAAA15468-0C CAAAA15468-0E CAAAA15468-36
Diethyl malonate 99%
Diethyl malonate

Purity: 99% CAS No.: 105-53-3 Molecular Formula: C7H12O4 Formula Weight: 160.17 Flash Point: 90°(194°F) Physical Form: Liquid Density: 1.055 Refractive Index: 1.4140 Odor: Pleasant Melting Point: -50° Boiling Point: 198-200° Beilstein Reference: 774687 Merck Reference: 14,3823 MDL No.: MFCD00009195

Formula: C₇H₁₂O₄
Boiling Pt: 198…200 °C
Melting Pt: –50 °C
Density: 1.055
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 14,3823

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR