4-Cyanobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A15434-06 A15434-14 A15434-22
CAAAA15434-06EA 66.68 CAD
CAAAA15434-06 CAAAA15434-14 CAAAA15434-22
4-Cyanobenzoic acid 98%
4-Cyanobenzoic acid

Purity: 98% CAS No.: 619-65-8 Molecular Formula: C8H5NO2 Formula Weight: 147.13 Melting Point: ca 221° dec. Beilstein Reference: 1862865

Formula: C₈H₅NO₂
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: ∼ 221 °C (dec.)
Density: 1.382 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002528
CAS Number: 619-65-8
EINECS: 210-606-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR