Europium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

15290-18 15290-04 15290-09
CAAAA15290-18EA 1474.14 CAD
CAAAA15290-18 CAAA15290-04 CAAA15290-09
Europium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Europium trinitrate hexahydrate

MDL: MFCD00149700


Purity: 99.9% CAS No.: 10031-53-5 Molecular Formula: Eu(NO3)3.6H2O Formula Weight: 446.06 (337.97anhy) Physical Form: Crystalline or powder Appearance: White Odor: Odorless Melting Point: 85° Merck Reference: 14,3904 UN No.: 1477 MDL No.: MFCD00149700

Formula: EuN₃O₉·6H₂O
Melting Pt: 85 °C
Density: 2.258 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149700
CAS Number: 10031-53-5
EINECS: 233-380-3
UN: 1477
ADR: 5.1,III
Merck Index: 14,3904

Order Now

Specification Test Results

Form Crystalline


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR