Pivaloyl chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

A15051-0F A15051-AE A15051-AP
CAAAA15051-0FEA 402.9 CAD
CAAAA15051-0F CAAAA15051-AE CAAAA15051-AP
Pivaloyl chloride 98+%
Pivaloyl chloride

Purity: 98+% CAS No.: 3282-30-2 Molecular Formula: (CH3)3CCOCl Formula Weight: 120.58 Flash Point: 8°(46°F) Refractive Index: 1.4120 Odor: Pungent Melting Point: -57° Boiling Point: 105-106° Beilstein Reference: 385668 UN No.: 2438 MDL No.: MFCD00000709

Formula: C₅H₉ClO
Boiling Pt: 105…106 °C
Melting Pt: –57 °C
Density: 0.985
Flash Pt: 8 °C (46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000709
CAS Number: 3282-30-2
EINECS: 221-921-6
UN: 2438
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR