3-Nitrobenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

A15040-14 A15040-22
CAAAAA15040-36EA 1421.65 CAD
CAAAAA15040-36 CAAAA15040-14 CAAAA15040-22
3-Nitrobenzyl alcohol 98%
3-Nitrobenzyl alcohol

Purity: 98% CAS No.: 619-25-0 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 110°(230°F) Refractive Index: 1.5755 Melting Point: 29-33° Boiling Point: 175-180°/3mm Beilstein Reference: 2044769 MDL No.: MFCD00007273

Formula: C₇H₇NO₃
Boiling Pt: 175…180 °C/3 mm
Melting Pt: 29…33 °C
Density: 1.300
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007273
CAS Number: 619-25-0
EINECS: 210-588-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR