D-(-)-Leucine 99%

Supplier: Thermo Scientific Chemicals

A14842-06
CAAAAA14842-22EA 1077.72 CAD
CAAAAA14842-22 CAAAA14842-06
D-(-)-Leucine 99%
D(-)-Leucine

Purity: 99% CAS No.: 328-38-1 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721721

Formula: C₆H₁₃NO₂
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00063088
CAS Number: 328-38-1
EINECS: 206-327-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR