Ethyl-6-chloronicotinate 97%

Supplier: Thermo Scientific Chemicals
Warning

A14723-06 A14723-14
CAAAA14723-06EA 59.25 CAD
CAAAA14723-06 CAAAA14723-14
Ethyl-6-chloronicotinate 97%
Ethyl 6-chloronicotinate

Purity: 97% CAS No.: 49608-01-7 Molecular Formula: C8H8ClNO2 Formula Weight: 185.61 Flash Point: 110°(230°F) Refractive Index: 1.5220 Melting Point: 26-30° Boiling Point: 86-87°/1mm Beilstein Reference: 4742175 MDL No.: MFCD00082739

Formula: C₇H₆ClNO₂
Boiling Pt: 86…87 °C/1 mm
Melting Pt: 26…30 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082739
CAS Number: 49608-01-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR