Phenylboronic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14257-09 A14257-18 A14257-30
CAAAA14257-09EA 65.89 CAD
CAAAA14257-09 CAAAA14257-18 CAAAA14257-30
Phenylboronic acid 98+%
Phenylboronic acid

Purity: 98+% CAS No.: 98-80-6 Molecular Formula: C6H7BO2 Formula Weight: 121.93 Melting Point: 214-219° Beilstein Reference: 970972 Merck Reference: 14,1068 MDL No.: MFCD00002103

Formula: C₆H₇BO₂
Melting Pt: 214…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00002103
CAS Number: 98-80-6
EINECS: 202-701-9
Merck Index: 14,1068

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR