1,3-Benzodioxole 99%

Supplier: Thermo Scientific Chemicals
Warning

A14192-0B A14192-18 A14192-30
CAAAA14192-0BEA 1091.78 CAD
CAAAA14192-0B CAAAA14192-18 CAAAA14192-30
1,3-Benzodioxole 99%
1,3-Benzodioxole

Purity: 99% CAS No.: 274-09-9 Molecular Formula: C7H6O2 Formula Weight: 122.12 Flash Point: 55°(131°F) Refractive Index: 1.5390 Odor: Aromatic Melting Point: -18° Boiling Point: 172-173° Beilstein Reference: 115506 MDL No.: MFCD00005818

Formula: C₇H₆O₂
Boiling Pt: 172…173 °C
Melting Pt: –18 °C
Density: 1.185
Flash Pt: 55 °C (131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005818
CAS Number: 274-09-9
EINECS: 205-992-0
UN: 1993
ADR: 3,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR