Butyl-4-hydroxybenzoate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben

A14043-30
CAAAA14043-0BEA 825.57 CAD
CAAAA14043-0B CAAAA14043-0I CAAAA14043-30
Butyl-4-hydroxybenzoate 99+%
Butyl 4-hydroxybenzoate

Purity: 99+% CAS No.: 94-26-8 Molecular Formula: C11H14O3 Formula Weight: 194.23 Melting Point: 67-71° Beilstein Reference: 1103741 Merck Reference: 14,1584 MDL No.: MFCD00016478

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 156…157 °C (3.5 mmHg)
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00016478
CAS Number: 94-26-8
EINECS: 202-318-7
Merck Index: 14,1584

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR