1-Bromo-3-chlorobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

A14026-22 A14026-36
CAAAA14026-22EA 121.66 CAD
CAAAA14026-22 CAAAA14026-36
1-Bromo-3-chlorobenzene 99%
1-Bromo-3-chlorobenzene

Purity: 99% CAS No.: 108-37-2 Molecular Formula: C6H4BrCl Formula Weight: 191.46 Flash Point: 80°(176°F) Refractive Index: 1.5770 Odor: Aromatic Melting Point: -22° Boiling Point: 195-196° Beilstein Reference: 1446353 MDL No.: MFCD00000568

Formula: C₆H₄BrCl
Boiling Pt: 195…196 °C
Melting Pt: –22 °C
Density: 1.630
Flash Pt: 80 °C(176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000568
CAS Number: 108-37-2
EINECS: 203-575-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR