Pyrazine, (max. 1% H₂O) 99+% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

A13982-14
CAAAA13982-14EA 106.81 CAD
CAAAA13982-14 CAAAA13982-22
Pyrazine, (max. 1% H₂O) 99+% (dry weight)
Pyrazine

Purity: 99+% CAS No.: 290-37-9 Molecular Formula: C4H4N2 Formula Weight: 80.09 Flash Point: 55°(131°F) Refractive Index: 1.5235 Odor: Strong Melting Point: 50-56° Boiling Point: 113-116° Beilstein Reference: 103905 Merck Reference: 14,7957 UN No.: 1325 MDL No.: MFCD00006122

Formula: C₄H₄N₂
Boiling Pt: 113…116 °C
Melting Pt: 50…56 °C
Density: 1.031
Flash Pt: 55 °C (131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006122
CAS Number: 290-37-9
EINECS: 206-027-6
UN: 1325
ADR: 4.1,III
Merck Index: 14,7957

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR