3-Methylthiophene-2-carboxaldehyde tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

A13961-14 A13961-22
CAAAA13961-14EA 65.41 CAD
CAAAA13961-14 CAAAA13961-22
3-Methylthiophene-2-carboxaldehyde tech. 90%, Technical Grade
3-Methylthiophene-2-carboxaldehyde

Purity: 90% CAS No.: 5834-16-2 Molecular Formula: C6H6OS Formula Weight: 126.18 Flash Point: 82°(179°F) Refractive Index: 1.5860 Boiling Point: 215° Beilstein Reference: 107874 MDL No.: MFCD00005430

Formula: C₆H₆OS
Boiling Pt: 215 °C
Density: 1.170
Flash Pt: 82 °C(179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005430
CAS Number: 5834-16-2
EINECS: 227-418-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR