Cyclopropylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger

A13844-09 A13844-18 A13844-30
CAAAA13844-09EA 69.36 CAD
CAAAA13844-09 CAAAA13844-18 CAAAA13844-30
Cyclopropylamine 98+%
Cyclopropylamine

Purity: 98+% CAS No.: 765-30-0 Molecular Formula: C3H7N Formula Weight: 57.10 Flash Point: -25°(-13°F) Refractive Index: 1.4206 Melting Point: -10° Boiling Point: 49-50° Beilstein Reference: 741858 UN No.: 2733 MDL No.: MFCD00001301

Formula: C₃H₇N
Boiling Pt: 49…50 °C
Melting Pt: –10 °C
Density: 0.824
Flash Pt: –25 °C (–13 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001301
CAS Number: 765-30-0
EINECS: 212-142-2
UN: 2733
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR