2',5'-Dichloroacetophenone 98%

Supplier: Thermo Scientific Chemicals

A13770-09 A13770-14 A13770-22
CAAAAA13770-09EA 120.24 CAD
CAAAAA13770-09 CAAAA13770-14 CAAAA13770-22
2',5'-Dichloroacetophenone 98%
2',5'-Dichloroacetophenone

Purity: 98% CAS No.: 2476-37-1 Molecular Formula: C8H6Cl2O Formula Weight: 189.04 Flash Point: 110°(230°F) Refractive Index: 1.5610 Melting Point: 11-13° Boiling Point: 118°/12mm Beilstein Reference: 1865043 MDL No.: MFCD00000607

Formula: C₈H₆Cl₂O
Boiling Pt: 118 °C/12 mm
Melting Pt: 11…13 °C
Density: 1.312
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000607
CAS Number: 2476-37-1
EINECS: 219-605-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR