N-Carbobenzyloxy-L-valine 99%

Supplier: Thermo Scientific Chemicals

A13583-14 A13583-22
CAAAA13583-06EA 56.37 CAD
CAAAA13583-06 CAAAA13583-14 CAAAA13583-22
N-Carbobenzyloxy-L-valine 99%
N-Carbobenzyloxy-L-valine

Purity: 99% CAS No.: 1149-26-4 Molecular Formula: C13H17NO4 Formula Weight: 251.28 Melting Point: 60° Beilstein Reference: 2056617 MDL No.: MFCD00008922

Formula: C13H17NO4
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008922
CAS Number: 1149-26-4
EINECS: 214-562-1

Order Now

Specification Test Results

Optical rotation -4.0° (c=2 in acetic acid)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR