1,4-Bis(2-methylstyryl)benzene 99%

Supplier: Thermo Scientific Chemicals

A13534-06 A13534-14
CAAAA13534-06EA 119.29 CAD
CAAAA13534-06 CAAAA13534-14
1,4-Bis(2-methylstyryl)benzene 99%
1,4-Bis(2-methylstyryl)benzene

Purity: 99% CAS No.: 13280-61-0 Molecular Formula: C24H22 Formula Weight: 310.44 Odor: Odorless Melting Point: 179-184° Beilstein Reference: 2375845 MDL No.: MFCD00008529

Formula: C₂₄H₂₂
Melting Pt: 179…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00008529
CAS Number: 13280-61-0
EINECS: 236-285-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR