4-Phenyl-1-butanol 97%

Supplier: Thermo Scientific Chemicals
Warning

A13433-09 A13433-18 A13433-30
CAAAA13433-09EA 148.05 CAD
CAAAA13433-09 CAAAA13433-18 CAAAA13433-30
4-Phenyl-1-butanol 97%
4-Phenyl-1-butanol

Purity: 97% CAS No.: 3360-41-6 Molecular Formula: C6H5(CH2)4OH Formula Weight: 150.22 Flash Point: 110°(230°F) Refractive Index: 1.5210 Boiling Point: 138-140°/14mm Beilstein Reference: 2042122 MDL No.: MFCD00002971

Formula: (C₆H₅)(CH₂)₃CH₂OH
Boiling Pt: 138…140 °C/14 mm
Density: 0.975
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002971
CAS Number: 3360-41-6
EINECS: 222-128-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR