Ethyl-4-nitrobenzoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 4-nitrobenzoate, 4-Nitrobenzoic acid ethyl ester

A13398-14 A13398-22 A13398-36
CAAAA13398-14EA 49.14 CAD
CAAAA13398-14 CAAAA13398-22 CAAAA13398-36
Ethyl-4-nitrobenzoate 98%
Ethyl 4-nitrobenzoate

Purity: 98% CAS No.: 99-77-4 Molecular Formula: C9H9NO4 Formula Weight: 195.18 Melting Point: 55-58° Beilstein Reference: 1912879 Merck Reference: 14,3832 MDL No.: MFCD00007351

Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 55…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00007351
CAS Number: 99-77-4
EINECS: 202-786-2
Merck Index: 14,3832

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR