Tributyltin methanolate 97%

Supplier: Thermo Scientific Chemicals
Danger

A13355-22 A13355-0B A13355-30
CAAAAA13355-22EA 376.04 CAD
CAAAAA13355-22 CAAAA13355-0B CAAAA13355-30
Tributyltin methanolate 97%
Tributyltin methanolate

Purity: 97% CAS No.: 1067-52-3 Molecular Formula: C13H30OSn Formula Weight: 321.09 Flash Point: 98°(208°F) Refractive Index: 1.4740 Boiling Point: 124-126°/4mm Beilstein Reference: 3600960 UN No.: 2788 MDL No.: MFCD00009419

Formula: C₁₃H₃₀OSn
Boiling Pt: 124…126 °C/4 mm
Density: 1.120
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009419
CAS Number: 1067-52-3
EINECS: 213-933-5
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR