3-Nitrophenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A13336-03 A13336-06 A13336-14
CAAAA13336-03EA 39.18 CAD
CAAAA13336-03 CAAAA13336-06 CAAAA13336-14
3-Nitrophenylboronic acid 98%
3-Nitrophenylboronic acid

Purity: 98% CAS No.: 13331-27-6 Molecular Formula: C6H6BNO4 Formula Weight: 166.93 Melting Point: ca 285° dec. Beilstein Reference: 2938638

Formula: C₆H₆BNO₄
Melting Pt: ∼ 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007193
CAS Number: 13331-27-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR