Methanesulfonic anhydride 98%

Supplier: Thermo Scientific Chemicals
Danger

A13328-14 A13328-09 A13328-18 A13328-30
CAAAAA13328-14EA 130.98 CAD
CAAAAA13328-14 CAAAA13328-09 CAAAA13328-18 CAAAA13328-30
Methanesulfonic anhydride 98%
Methanesulphonic anhydride

Purity: 98% CAS No.: 7143-01-3 Molecular Formula: C2H6O5S2 Formula Weight: 174.20 Melting Point: 65-69° Boiling Point: 125°/4mm UN No.: 3261 MDL No.: MFCD00007556

Formula: C₂H₆O₅S₂
Boiling Pt: 125 °C/4 mm
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00007556
CAS Number: 7143-01-3
EINECS: 230-442-1
UN: 3261
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR