1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

A13190-03 A13190-06 A13190-14
CAAAA13190-03EA 255.96 CAD
CAAAA13190-03 CAAAA13190-06 CAAAA13190-14
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid

Acts as a calcium chelator


Purity: 97% CAS No.: 85233-19-8 Molecular Formula: C22H24N2O10 Formula Weight: 476.44 Storage Temperature: 2°C to 8°C Melting Point: ca 174° dec. MDL No.: MFCD00036255

Formula: C₂₂H₂₄N₂O₁₀
Melting Pt: ca 174 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00036255
CAS Number: 85233-19-8
Merck Index: 14,957

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR