2-Aminobenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

A13180-09 A13180-18 A13180-30
CAAAA13180-09EA 52.77 CAD
CAAAA13180-09 CAAAA13180-18 CAAAA13180-30
2-Aminobenzyl alcohol 98%
2-Aminobenzyl alcohol

Purity: 98% CAS No.: 5344-90-1 Molecular Formula: C7H9NO Formula Weight: 123.16 Storage Temperature: 2°C to 8°C Melting Point: 81-84° Boiling Point: 160°/10mm Beilstein Reference: 1072211 MDL No.: MFCD00007749

Formula: C₇H₉NO
Boiling Pt: 160 °C/10 mm
Melting Pt: 81…84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007749
CAS Number: 5344-90-1
EINECS: 226-293-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR