DL-Ethionine 98%

Supplier: Thermo Scientific Chemicals
Warning

A13154-03 A13154-06 A13154-14
CAAAA13154-03EA 82.63 CAD
CAAAA13154-03 CAAAA13154-06 CAAAA13154-14
DL-Ethionine 98%
DL-Ethionine

Purity: 98% CAS No.: 67-21-0 Molecular Formula: C6H13NO2S Formula Weight: 163.24 Odor: Odorless Melting Point: ca 265° dec. Beilstein Reference: 1722529 Merck Reference: 14,3738 MDL No.: MFCD00063102

Formula: C₆H₁₃NO₂S
Melting Pt: ∼ 265 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00063102
CAS Number: 67-21-0
EINECS: 200-647-0
Merck Index: 14,3738

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR