L(+)-Lysine monohydrate 97%

Supplier: Thermo Scientific Chemicals

A13040-14 A13040-22 A13040-36
CAAAA13040-14EA 94.33 CAD
CAAAA13040-14 CAAAA13040-22 CAAAA13040-36
L(+)-Lysine monohydrate 97%
L(+)-Lysine monohydrate

Purity: 97% CAS No.: 39665-12-8 Molecular Formula: C6H14N2O2.H2O Formula Weight: 164.21 (146.19anhy) Melting Point: 210° dec. Beilstein Reference: 1722531 Merck Reference: 14,5636 Optical Rotation: +22.5° (c=2 in 6M HCl) MDL No.: MFCD00151035

Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151035
CAS Number: 39665-12-8
EINECS: 200-294-2
Merck Index: 14,5636

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR