Iodotrimethylsilane 97% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with copper

A12902-06 A12902-14 A12902-22 A12902-30
CAAAAA12902-06EA 72.52 CAD
CAAAAA12902-06 CAAAAA12902-14 CAAAAA12902-22 CAAAA12902-30
Iodotrimethylsilane 97% stabilized
Iodotrimethylsilane

Purity: 97% CAS No.: 16029-98-4 Molecular Formula: C3H9ISi Formula Weight: 200.09 Storage Temperature: -30°C to -10°C Flash Point: -2°(28°F) Refractive Index: 1.4760 Boiling Point: 105-106° Beilstein Reference: 1731136 UN No.: 2924 MDL No.: MFCD00001028

Formula: C₃H₉ISi
Boiling Pt: 105…106 °C
Melting Pt: < 0 °C
Density: 1.440
Flash Pt: –2 °C (28 °F)
Storage Temperature: Freezer
MDL Number: MFCD00001028
CAS Number: 16029-98-4
EINECS: 240-171-0
UN: 2924
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR