4-Chloroaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

A12879-0B A12879-0I A12879-30
CAAAA12879-0BEA 188.02 CAD
CAAAA12879-0B CAAAA12879-0I CAAAA12879-30
4-Chloroaniline 98%
4-Chloroaniline

Purity: 98% CAS No.: 106-47-8 Molecular Formula: C6H6ClN Formula Weight: 127.57 Flash Point: 120°(248°F) Melting Point: 67-72° Boiling Point: 232° Beilstein Reference: 471359 Merck Reference: 14,2118 UN No.: 2018 MDL No.: MFCD00007835

Formula: C₆H₆ClN
Boiling Pt: 232 °C
Melting Pt: 67…72 °C
Density: 1.430
Flash Pt: 120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007835
CAS Number: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1,II
Merck Index: 14,2118

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR