Procion blue H-5R

Supplier: Thermo Scientific Chemicals

A12719-14 A12719-22
CAAAA12719-14EA 71.42 CAD
CAAAA12719-14 CAAAA12719-22
Procion blue H-5R
Procion blue H-5R

CAS No.: 12236-84-9 MDL No.: MFCD00070851

Storage Temperature: Ambient
MDL Number: MFCD00070851
CAS Number: 12236-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR