(S)-(-)-α-Methylbenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

A12632-14 A12632-22 A12632-36
CAAAA12632-14EA 91.32 CAD
CAAAA12632-14 CAAAA12632-22 CAAAA12632-36
(S)-(-)-α-Methylbenzylamine 98%
(S)-(-)-α-Methylbenzylamine

Purity: 98% CAS No.: 2627-86-3 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 71°(159°F) Refractive Index: 1.5260 Odor: Amine-like Melting Point: -10° Boiling Point: 187° Beilstein Reference: 2204907 Merck Reference: 14,6026 Optical Rotation: -30° (c=10 in ethanol) UN No.: 2735 MDL No.: MFCD00064406

Formula: C₈H₁₁N
Boiling Pt: 187 °C
Melting Pt: –10 °C
Density: 0.948
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064406
CAS Number: 2627-86-3
EINECS: 220-098-0
UN: 2735
ADR: 8,III
Merck Index: 14,6026

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR