Ethyl-4-chlorobenzoate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-chlorobenzoate, 4-Chlorobenzoic acid ethyl ester

A12540-18 A12540-30
CAAAA12540-0BEA 1052.96 CAD
CAAAA12540-0B CAAAA12540-18 CAAAA12540-30
Ethyl-4-chlorobenzoate 98+%
Ethyl 4-chlorobenzoate

Purity: 98+% CAS No.: 7335-27-5 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Refractive Index: 1.5240 Boiling Point: 237-239° MDL No.: MFCD00013645

Formula: C₉H₉ClO₂
MW: 184.62
Boiling Pt: 237…239 °C
Density: 1.18
Storage Temperature: Ambient
MDL Number: MFCD00013645
CAS Number: 7335-27-5
EINECS: 230-844-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR