3-Nitro-p-toluidine 98%

Supplier: Thermo Scientific Chemicals
Warning

A12538-14 A12538-22
CAAAA12538-14EA 115.34 CAD
CAAAA12538-14 CAAAA12538-22
3-Nitro-p-toluidine 98%
3-Nitro-p-toluidine

Purity: 98% CAS No.: 119-32-4 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Flash Point: 175°(347°F) Odor: Characteristic Beilstein Reference: 638984 UN No.: 2660 MDL No.: MFCD00007910

Formula: C₇H₈N₂O₂
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 74…79 °C
Density: 1.312
Flash Pt: 175 °C (347 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007910
CAS Number: 119-32-4
EINECS: 204-314-0
UN: 2660
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR