Trimethylsilyl trifluoromethanesulfonate 99%

Supplier: Thermo Scientific Chemicals
Danger

A12535-09 A12535-18 A12535-30
CAAAA12535-09EA 73.15 CAD
CAAAA12535-09 CAAAA12535-18 CAAAA12535-30
Trimethylsilyl trifluoromethanesulfonate 99%
Trimethylsilyl trifluoromethanesulphonate

Purity: 99% CAS No.: 27607-77-8 Molecular Formula: (CH3)3SiSO3CF3 Formula Weight: 222.26 Flash Point: 25°(77°F) Refractive Index: 1.3600 Boiling Point: 39-40°/12mm Beilstein Reference: 1868911 Merck Reference: 14,9719 UN No.: 2924 MDL No.: MFCD00000406

Formula: C₄H₉F₃O₃SSi
Boiling Pt: 39…40 °C/12 mm
Density: 1.225
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000406
CAS Number: 27607-77-8
EINECS: 248-565-4
UN: 2920
ADR: 8,II
Merck Index: 14,9719

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR