Phenethyl bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

A12528-22 A12528-36
CAAAA12528-22EA 57.83 CAD
CAAAA12528-22 CAAAA12528-36
Phenethyl bromide 98%
Phenethyl bromide

Purity: 98% CAS No.: 103-63-9 Molecular Formula: C8H9Br Formula Weight: 185.06 Flash Point: 89°(192°F) Refractive Index: 1.5560 Melting Point: -56° Boiling Point: 220-221° Beilstein Reference: 507487 MDL No.: MFCD00000240

Formula: BrCH₂CH₂C₆H₅
Boiling Pt: 220…221 °C
Melting Pt: –56 °C
Density: 1,355
Flash Pt: 89 °C (192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000240
CAS Number: 103-63-9
EINECS: 203-130-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR