4-(Methylamino)benzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

A12426-06 A12426-14 A12426-22
CAAAA12426-06EA 57.04 CAD
CAAAA12426-06 CAAAA12426-14 CAAAA12426-22
4-(Methylamino)benzoic acid 97%
4-(Methylamino)benzoic acid

Purity: 97% CAS No.: 10541-83-0 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 158-160° Beilstein Reference: 2082805

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 296
Melting Pt: 159
Storage Temperature: Ambient
MDL Number: MFCD00002535
CAS Number: 10541-83-0
EINECS: 234-121-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR